(R)-(-)-Nipecotic acid ≥96%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(R)-Piperidine-3-carboxylic acid, (R)-(-)-3-Piperidinecarboxylic acid, D-Nipecotic acid
H57434.03
H57434.06
H57434.14
H57434.03EA
1540
CZK
H57434.03
H57434.06
H57434.14
(R)-(-)-Nipecotic acid ≥96%
(R)-(-)-Nipecotic acid
Purity: 96+%
CAS No.: 25137-00-2
Molecular Formula: C6H11NO2
Formula Weight: 129.16
Melting Point: 250-255°
Optical Rotation: -5° (c=1 in water)
Vzorec:
C₆H₁₁NO₂ Bod tání: 250…255 °C Storage Temperature: Ambient |
MDL Number:
MFCD01630787 CAS číslo: 25137-00-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...