Tisk…

(S)-(-)-4-Amino-2-hydroxybutyric acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (S)-4-Amino-2-hydroxybutanoic acid

M-Clarity™ quality level = MQ100

467359-25G 467359-5G
SIAL467359-25GEA 3920 CZK
SIAL467359-25G SIAL467359-5G
(S)-(-)-4-Amino-2-hydroxybutyric acid, Sigma-Aldrich®
(S)-(-)-4-Amino-2-hydroxybutyric acid
Vzorec: C₄H₉NO₃
MDL Number: MFCD00674110
CAS číslo: 40371-51-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR