Tisk…

(S)-(-)-4-Amino-2-hydroxybutyric acid 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: (S)-4-Amino-2-hydroxybutanoic acid

H28796.06 H28796.14
H28796.06EA 1740 CZK
H28796.06 H28796.14
(S)-(-)-4-Amino-2-hydroxybutyric acid 98%
(S)-(-)-4-Amino-2-hydroxybutyric acid

Purity: 98% CAS No.: 40371-51-5 Molecular Formula: C4H9NO3 Formula Weight: 119.12 MDL No.: MFCD00674110

Vzorec: C₄H₉NO₃
MW: 119,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00674110
CAS číslo: 40371-51-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR