Tisk…

2-Coumaranone ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: Benzofuran-2(3H)-one, Benzo[b]furan-2(3H)-one

A18617.03 A18617.06 A18617.14
A18617.03EA 1050 CZK
A18617.03 A18617.06 A18617.14
2-Coumaranone ≥99%
2-Coumaranone

Purity: 99% CAS No.: 553-86-6 Molecular Formula: C8H6O2 Formula Weight: 134.13 Flash Point: 110°(230°F) Odour: Characteristic Boiling Point: 248-250° Beilstein Reference: 114689 MDL No.: MFCD00005856

Vzorec: C₈H₆O₂
MW: 134,13 g/mol
Bod varu: 248…250 °C
Bod tání: 47…52 °C
Hustota: 1,22
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005856
CAS číslo: 553-86-6
EINECS: 209-052-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR