Tisk…

2-Nitro-5-(trifluoromethyl)aniline ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L12310.03 L12310.06
L12310.03EA 2570 CZK
L12310.03 L12310.06
2-Nitro-5-(trifluoromethyl)aniline ≥97%
2-Nitro-5-(trifluoromethyl)aniline

Purity: 97% CAS No.: 402-14-2 Molecular Formula: C7H5F3N2O2 Formula Weight: 206.13 Melting Point: 98-102° Beilstein Reference: 2732344 UN No.: 2811 MDL No.: MFCD00042447

Vzorec: C₇H₅F₃N₂O₂
Bod tání: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00042447
CAS číslo: 402-14-2
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR