Tisk…

2-Fluoro-4-(trifluoromethyl)phenylboronic acid ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Fluoro-4-(trifluoromethyl)benzeneboronic acid

H28788.06 H28788.03
H28788.06EA 11360 CZK
H28788.06 H28788.03
2-Fluoro-4-(trifluoromethyl)phenylboronic acid ≥97%
2-Fluoro-4-(trifluoromethyl)phenylboronic acid

Purity: 97% CAS No.: 503309-11-3 Molecular Formula: C7H5BF4O2 Formula Weight: 207.92 Melting Point: 113-115° MDL No.: MFCD03094995

Vzorec: C₇H₅BF₄O₂
MW: 207,92 g/mol
Bod tání: 113…115 °C
Storage Temperature: Ambient
MDL Number: MFCD03094995
CAS číslo: 503309-11-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR