Tisk…

(4-(Hydroxymethyl)phenyl)acetic acid ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-(4-(Hydroxymethyl)phenyl)acetic acid, 4-(Hydroxymethyl)phenylacetic acid

H27531.06 H27531.03
H27531.06EA 4230 CZK
H27531.06 H27531.03
(4-(Hydroxymethyl)phenyl)acetic acid ≥98%
(4-(Hydroxymethyl)phenyl)acetic acid

Purity: 98% CAS No.: 73401-74-8 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 131-134° MDL No.: MFCD00065692

Vzorec: C₉H₁₀O₃
MW: 166,18 g/mol
Bod tání: 131…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00065692
CAS číslo: 73401-74-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR