Tisk…

4-(Ethylthio)benzaldehyde ≥97%

Dodavatel: Thermo Fisher Scientific

L17624.03 L17624.06
L17624.03EA 1740 CZK
L17624.03 L17624.06
4-(Ethylthio)benzaldehyde ≥97%
4-(Ethylthio)benzaldehyde

Purity: 98% CAS No.: 84211-94-9 Molecular Formula: C9H10OS Formula Weight: 166.24 Refractive Index: 1.6240 Boiling Point: 151-153°/15mm MDL No.: MFCD01318145

Vzorec: C₉H₁₀OS
Bod varu: 151…153 °C (15 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD01318145
CAS číslo: 84211-94-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR