Pentafluorophenyl isothiocyanate, Sigma-Aldrich®
Dodavatel: Merck
76755-1ML
76755-5ML
SIAL76755-1MLEA
5800
CZK
SIAL76755-1ML
SIAL76755-5ML
Pentafluorophenyl isothiocyanate, Sigma-Aldrich®
Pentafluorophenyl isothiocyanate
Vzorec:
C₇F₅NS MW: 225.14 g/mol |
MDL Number:
MFCD00041042 CAS číslo: 35923-79-6 EINECS: 252-791-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...