4-Benzoyl-4'-bromobiphenyl ≥99%
Dodavatel: Thermo Fisher Scientific
A18482.14
A18482.22
A18482.14EA
1890
CZK
A18482.14
A18482.22
4-Benzoyl-4'-bromobiphenyl ≥99%
4-Benzoyl-4'-bromobiphenyl
Purity: 99%
CAS No.: 63242-14-8
Molecular Formula: C19H13BrO
Formula Weight: 337.22
Melting Point: 155-158°
Beilstein Reference: 2378429
Vzorec:
C₁₉H₁₃BrO Bod tání: 155…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00209630 CAS číslo: 63242-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...