Tisk…

Tetrabutylammonium dihydrogen phosphate ≥98%, non-UV

Dodavatel: Thermo Scientific
Varování

Synonyms: Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate

46746.04 46746.09 46746.18
46746.04EA 2820 CZK
46746.04 46746.09 46746.18
Tetrabutylammonium dihydrogen phosphate ≥98%, non-UV
Tetrabutylammonium dihydrogen phosphate

Tetra-n-butylammonium phosphate, is used in chemicals, pharmaceutical research, pharmaceutical intermediates, and reagents. Tetrabutylammonium phosphate can be used in organic synthesis. It is used as an Ion-associating reagent for HPLC.

Vzorec: C₁₆H₃₈NO₄P
MW: 339,46 g/mol
Bod tání: 151…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00064526
CAS číslo: 5574-97-0
EINECS: 226-947-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR