Diphosphoryl tetrachloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Diphosphoryl chloride, Pyrophosphoryl chloride
M-Clarity™ quality level = MQ100
381829-25ML
381829-5ML
SIAL381829-25MLEA
23120
CZK
SIAL381829-25ML
SIAL381829-5ML
Diphosphoryl tetrachloride, Sigma-Aldrich®
Diphosphoryl tetrachloride
Vzorec:
Cl₄O₃P₂ MW: 251,76 g/mol |
MDL Number:
MFCD00061542 CAS číslo: 13498-14-1 EINECS: 236-824-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...