Tisk…

4-(Tributylstannyl)pyridine 96%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4-(Tri-n-butylstannyl)pyridine

H51435.03 H51435.06
H51435.03EA 3210 CZK
H51435.03 H51435.06
4-(Tributylstannyl)pyridine 96%
4-(Tributylstannyl)pyridine

Purity: 96% CAS No.: 124252-41-1 Molecular Formula: C17H31NSn Formula Weight: 368.15 UN No.: 2788 MDL No.: MFCD01319071

Vzorec: C₁₇H₃₁NSn
MW: 368,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01319071
CAS číslo: 124252-41-1
UN: 2788
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR