Tisk…

Chloraceton 95% stabilizovaný

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Chloropropanone

Stabilization: Stabilised with epoxidized soybean oil

A11922.0B A11922.30
A11922.0BEA 4400 CZK
A11922.0B A11922.30
Chloraceton 95% stabilizovaný
Chloraceton

Stabilized with 0.1% epoxidized soybean oil


Purity: 95% CAS No.: 78-95-5 Molecular Formula: C3H5ClO Formula Weight: 92.53 Storage Temperature: 2°C to 8°C Flash Point: 40°(104°F) Physical Form: Colourless to brown liquid Density: 1.161 Refractive Index: 1.4340 Appearance: Colourless to brown Odour: Pungent pH: 4.3 Melting Point: -45° Boiling Point: 118-120° Beilstein Reference: 605369 Merck Reference: 14,2114 UN No.: 1695 MDL No.: MFCD00000936

Vzorec: C₃H₅ClO
MW: 92,52 g/mol
Bod varu: 118…120 °C
Bod tání: –45 °C
Hustota: 1,161
Bod vzplanutí: 40 °C (104 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000936
CAS číslo: 78-95-5
EINECS: 201-161-1
UN: 1695
ADR: 6.1,I
Merck Index: 14,2114

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR