5-Methylphenazinium methyl sulphate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
PMS, 5-Methylphenazin-5-ium methyl sulphate, Phenazine methosulphate, Phenazine methylsulphate
H56718.06
H56718.14
H56718.06EA
6010
CZK
H56718.06
H56718.14
5-Methylphenazinium methyl sulphate ≥98%
5-Methylphenazinium methyl sulphate
Purity: 98+%
CAS No.: 299-11-6
Molecular Formula: C13H11N2.CH3SO4
Formula Weight: 306.34
Storage Temperature: -30°C to -10°C
Vzorec:
C₁₄H₁₄N₂O₄S MW: 306,34 g/mol Bod tání: 198 °C Storage Temperature: Freezer |
MDL Number:
MFCD00011923 CAS číslo: 299-11-6 EINECS: 206-072-1 Merck Index: 12,06185 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...