Tisk…

Ethynylcyklopropan ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Cyclopropylacetylene

H53440.06 H53440.14 H53440.22
H53440.06EA 1630 CZK
H53440.06 H53440.14 H53440.22
Ethynylcyklopropan ≥97%
Ethynylcyklopropan

Purity: 97% CAS No.: 6746-94-7 Molecular Formula: C5H6 Formula Weight: 66.10 Flash Point: -17°(-1°F) Refractive Index: 1.429 Boiling Point: 51° UN No.: 3295 MDL No.: MFCD02181090

Vzorec: C₅H₆
Bod varu: 51 °C
Hustota: 0,781
Bod vzplanutí: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD02181090
CAS číslo: 6746-94-7
UN: 3295
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR