3-Chloro-5-methoxybenzoic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H31960.03
H31960.06
H31960.03EA
707
CZK
H31960.03
H31960.06
3-Chloro-5-methoxybenzoic acid ≥98%
3-Chloro-5-methoxybenzoic acid
Purity: 98%
CAS No.: 82477-67-6
Molecular Formula: C8H7ClO3
Formula Weight: 186.59
MDL No.: MFCD01317994
Vzorec:
C₈H₇ClO₃ Storage Temperature: Ambient |
MDL Number:
MFCD01317994 CAS číslo: 82477-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...