Tisk…

4-Phenyl-3H-1,2,4-triazole-3,5-(4H)-dione, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3,5-Dihydro-4-phenyl-4H-1,2,4-triazole-3,5-dione, PTAD, 4-Phenyl-1,2,4-triazoline-3,5-dione, 4-Phenyl-3H-1,2,4-triazole-3,5(4H)-dione

M-Clarity™ quality level = MQ100

280992-1G 280992-5G
SIAL280992-1GEA 3510 CZK
SIAL280992-1G SIAL280992-5G
4-Phenyl-3H-1,2,4-triazole-3,5-(4H)-dione, Sigma-Aldrich®
4-Phenyl-3H-1,2,4-triazole-3,5-(4H)-dione
Vzorec: C₈H₅N₃O₂
MDL Number: MFCD00003148
CAS číslo: 4233-33-4
EINECS: 224-191-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR