2-(Hydroxymethyl)-18-crown-6, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
388440-1G
388440-250MG
SIAL388440-1GEA
6050
CZK
SIAL388440-1G
SIAL388440-250MG
2-(Hydroxymethyl)-18-crown-6, Sigma-Aldrich®
2-(Hydroxymethyl)-18-crown-6
Vzorec:
C₁₃H₂₆O₇ |
MDL Number:
MFCD00188046 CAS číslo: 70069-04-4 EINECS: 274-299-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...