Tisk…

Coumalic acid ≥97%

Dodavatel: Thermo Fisher Scientific

B25521.06 B25521.14
B25521.06EA 810 CZK
B25521.06 B25521.14
Coumalic acid ≥97%
Coumalic acid

Purity: 97% CAS No.: 500-05-0 Molecular Formula: C6H4O4 Formula Weight: 140.10 Melting Point: ca 208° dec. Boiling Point: 218°/120mm Merck Reference: 14,2557 MDL No.: MFCD00006644

Vzorec: C₆H₄O₄
Bod varu: 218 °C (120 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00006644
CAS číslo: 500-05-0
EINECS: 207-899-0
Merck Index: 14,2557

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR