Tisk…

Methyl coumalate ≥98%

Dodavatel: Thermo Scientific

Synonyms: Methyl 2-oxo-2H-pyran-5-carboxylate

B25330.04 B25330.09
B25330.04EA 1770 CZK
B25330.04 B25330.09
Methyl coumalate ≥98%
Methyl coumalate

Purity: 98% CAS No.: 6018-41-3 Molecular Formula: C7H6O4 Formula Weight: 154.12 Melting Point: 68-72° Boiling Point: 178-180°/60mm Merck Reference: 14,2557 MDL No.: MFCD00010120

Vzorec: C₇H₆O₄
MW: 154,12 g/mol
Bod varu: 178…180 °C (60 mmHg)
Bod tání: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00010120
CAS číslo: 6018-41-3
EINECS: 227-871-1
Merck Index: 14,2557

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR