(Benzoylmethylene)triphenylphosphorane ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-Phenyl-2-(triphenylphosphoranylidene)ethanone
A12184.06
A12184.14
A12184.22
A12184.06EA
2470
CZK
A12184.06
A12184.14
A12184.22
(Benzoylmethylene)triphenylphosphorane ≥98%
(Benzoylmethylene)triphenylphosphorane
Purity: 98+%
CAS No.: 859-65-4
Molecular Formula: C26H21OP
Formula Weight: 380.43
Storage Temperature: 2°C to 8°C
Melting Point: 183-186°
Beilstein Reference: 620285
MDL No.: MFCD00014088
Vzorec:
C₂₆H₂₁OP MW: 380,43 g/mol Bod tání: 183…186 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00014088 CAS číslo: 859-65-4 EINECS: 212-727-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...