Tisk…

α,α-Diphenyl-N-methyl-D-prolinol, Sigma-Aldrich®

Dodavatel: Merck

SIAL43118-250MGEA 2050 CZK
SIAL43118-250MG
α,α-Diphenyl-N-methyl-D-prolinol, Sigma-Aldrich®
α,α-Diphenyl-N-methyl-D-prolinol
Vzorec: C₁₈H₂₁NO
MDL Number: MFCD00145217
CAS číslo: 144119-12-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR