Tisk…

N-Boc-L-prolinol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N-(tert-Butoxycarbonyl)-L-prolinol, 1-Boc-2-(S)-pyrrolidinemethanol, (S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (S)-1-Boc-2-pyrrolidinemethanol, N-Boc-L-Prolinol

M-Clarity™ quality level = MQ100

446327-1G 446327-5G
SIAL446327-1GEA 1550 CZK
SIAL446327-1G SIAL446327-5G
N-Boc-L-prolinol, Sigma-Aldrich®
N-Boc-L-prolinol
Vzorec: C₁₀H₁₉NO₃
MW: 201,27 g/mol
MDL Number: MFCD00066232
CAS číslo: 69610-40-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR