Tisk…

N-Boc-L-prolinol ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: N-(tert-Butoxycarbonyl)-L-prolinol, 1-Boc-2-(S)-pyrrolidinemethanol, (S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (S)-1-Boc-2-pyrrolidinemethanol, N-Boc-L-Prolinol

L09885.03 L09885.06 L09885.14
L09885.03EA 1240 CZK
L09885.03 L09885.06 L09885.14
N-Boc-L-prolinol ≥98%
N-Boc-L-prolinol

Purity: 98+% CAS No.: 69610-40-8 Molecular Formula: C10H19NO3 Formula Weight: 201.27 Storage Temperature: 2°C to 8°C Melting Point: 59-62° Beilstein Reference: 3542667 Optical Rotation: -54° (c=5 in methanol) MDL No.: MFCD00066232

Vzorec: C₁₀H₁₉NO₃
Bod tání: 59…62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066232
CAS číslo: 69610-40-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR