N-Boc-L-prolinol 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
N-(tert-Butoxycarbonyl)-L-prolinol, 1-Boc-2-(S)-pyrrolidinemethanol, (S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (S)-1-Boc-2-pyrrolidinemethanol, N-Boc-L-Prolinol
344560050
344560250
ACRO344560050EA
2270
CZK
ACRO344560050
ACRO344560250
N-Boc-L-prolinol 98%
N-Boc-L-prolinol
Vzorec:
C₁₀H₁₉NO₃ MW: 201,27 g/mol Bod tání: 60…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066232 CAS číslo: 69610-40-8 |
Výsledky specifikačního testu
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 58 to 63 °C |
GC | ≥97.5 % |
Specific optical rotation | -46° to -52° (20 °C, 589 nm) (c=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...