(±)-10-HDoHE
Dodavatel: Cayman Chemical
33400-25
33400-50
33400-100
33400-250
CAYM33400-25EA
3160
CZK
CAYM33400-25
CAYM33400-50
CAYM33400-100
CAYM33400-250
(±)-10-HDoHE
(±)-10-HDoHE
Vzorec:
C₂₂H₃₂O₃ |
CAS číslo:
90780-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...