Tisk…
Dicyclohexano-18-crown-6 (mixture of isomers) ≥97%
Dodavatel: Thermo Fisher Scientific
Nebezpečí
A15344.03 A15344.09 A15344.14
A15344.03EA 1060 CZK
A15344.03 A15344.09 A15344.14
Dicyclohexano-18-crown-6 (mixture of isomers) ≥97%
Dicyclohexano-18-crown-6 (mixture of isomers)
Purity: 97% CAS No.: 16069-36-6 Molecular Formula: C20H36O6 Formula Weight: 372.50 Flash Point: 110°(230°F) Odour: Odourless Beilstein Reference: 1130529 Merck Reference: 14,2602 UN No.: 2811 MDL No.: MFCD00005099
Vzorec: C₂₀H₃₆O₆
MW: 372.51 g/mol
Bod tání: 46…53 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005099
CAS číslo: 16069-36-6
EINECS: 240-216-4
UN: 2811
ADR: 6.1 III
Merck Index: 14,2602