Tisk…

Sulphur trioxide trimethylamine complex, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

135879-100G 135879-500G 135879-5G
SIAL135879-100GEA 3500 CZK
SIAL135879-100G SIAL135879-500G SIAL135879-5G
Sulphur trioxide trimethylamine complex, Sigma-Aldrich®
Sulphur trioxide trimethylamine complex
Vzorec: C₃H₉NO₃S
MW: 139.17 g/mol
MDL Number: MFCD00012421
CAS číslo: 3162-58-1
EINECS: 221-614-7
UN: 3261
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR