2-Hydroxypyridine-N-oxide ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Pyridinol-1-oxide
H54207.06
H54207.14
H54207.22
H54207.06EA
948
CZK
H54207.06
H54207.14
H54207.22
2-Hydroxypyridine-N-oxide ≥98%
2-Hydroxypyridine-N-oxide
Purity: 98+%
CAS No.: 13161-30-3
Molecular Formula: C5H5NO2
Formula Weight: 111.10
Flash Point: 100°(212°F)
Melting Point: 147-152°
Beilstein Reference: 1422545
MDL No.: MFCD00006195
Vzorec:
C₅H₅NO₂ MW: 111,10 g/mol Bod tání: 147…152 °C Hustota: 1,111 Bod vzplanutí: 100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006195 CAS číslo: 13161-30-3 EINECS: 236-100-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...