Tisk…

Hydroxylamine-O-sulphonic acid 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A12560.14 A12560.22
A12560.14EA 2130 CZK
A12560.14 A12560.22
Hydroxylamine-O-sulphonic acid 97%
Hydroxylamine-O-sulphonic acid

Purity: 97% CAS No.: 2950-43-8 Molecular Formula: NH2OSO3H Formula Weight: 113.09 Storage Temperature: 2°C to 8°C Melting Point: ca 210° dec. Beilstein Reference: 956566 UN No.: 3260 MDL No.: MFCD00011604

Vzorec: H₃NO₄S
MDL Number: MFCD00011604
CAS číslo: 2950-43-8
EINECS: 220-971-6
UN: 3260
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR