Triallyl phosphite
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
17484.09
17484.18
17484.30
17484.09EA
2730
CZK
17484.09
17484.18
17484.30
Triallyl phosphite
Triallyl phosphite
CAS No.: 102-84-1
Molecular Formula: C9H15O3P
Formula Weight: 202.19
Flash Point: 86°(187°F)
Physical Form: Liquid
Sensitivity: Air and Moisture Sensitive
Appearance: Colourless
Odour: Strong
Boiling Point: 84°/12mm
Beilstein Reference: 1705535
UN No.: 3278
MDL No.: MFCD00014948
Vzorec:
C₉H₁₅O₃P Bod varu: 84 °C (12 mmHg) Bod vzplanutí: 86 °C (187 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014948 CAS číslo: 102-84-1 EINECS: 203-060-8 UN: 3278 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...