Tisk…

Methyl dichlorophosphite, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl phosphorodichloridite

M-Clarity™ quality level = MQ100

235229-10G 235229-50G
SIAL235229-10GEA 1850 CZK
SIAL235229-10G SIAL235229-50G
Methyl dichlorophosphite, Sigma-Aldrich®
Methyl dichlorophosphite
Vzorec: CH₃Cl₂OP
MW: 132.91 g/mol
MDL Number: MFCD00000524
CAS číslo: 3279-26-3
EINECS: 221-915-3
UN: 2920
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR