Tisk…
O-Methylhydroxylamine hydrochloride ≥98%
Dodavatel: Thermo Fisher Scientific
Nebezpečí
Synonyms: Methylhydroxylamine hydrochloride, O-Methylhydroxylammonium chloride, Methoxyamine hydrochloride, Methoxylamine hydrochloride
A19188.06 A19188.14
A19188.06EA 3000 CZK
A19188.06 A19188.14
O-Methylhydroxylamine hydrochloride ≥98%
O-Methylhydroxylamine hydrochloride
Purity: 98+% CAS No.: 593-56-6 Molecular Formula: CH3ONH2.HCl Formula Weight: 83.52 Melting Point: ca 148° dec. Beilstein Reference: 3589723 Merck Reference: 14,5989 UN No.: 3261 MDL No.: MFCD00012951
Vzorec: CH₆ClNO
MW: 83.515 g/mol
Bod tání: ∼ 148 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00012951
CAS číslo: 593-56-6
EINECS: 209-798-7
UN: 3261
ADR: 8 III