Tisk…

O-(4-Nitrobenzyl)hydroxylamine hydrochloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: O-(4-Nitrobenzyl)-hydroxylammonium chloride

73200-1G 73200-5G
SIAL73200-1GEA 2340 CZK
SIAL73200-1G SIAL73200-5G
O-(4-Nitrobenzyl)hydroxylamine hydrochloride, Sigma-Aldrich®
O-(4-Nitrobenzyl)hydroxylamine hydrochloride
Vzorec: C₇H₉ClN₂O₃
MW: 204,61 g/mol
MDL Number: MFCD00012954
CAS číslo: 2086-26-2
EINECS: 218-228-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR