Tisk…

2,6-Xylyl isocyanide, Sigma-Aldrich®

Dodavatel: Merck

41413-25G 41413-5G
SIAL41413-25GEA 39430 CZK
SIAL41413-25G SIAL41413-5G
2,6-Xylyl isocyanide, Sigma-Aldrich®
2,6-Xylyl isocyanide
Vzorec: C₉H₉N
MW: 131,18 g/mol
MDL Number: MFCD00013479
CAS číslo: 2769-71-3
EINECS: 220-459-2
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR