Tisk…

Triphenylmethyl mercaptan, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

T84107-100G T84107-25G T84107-5G
SIALT84107-100GEA 11720 CZK
SIALT84107-100G SIALT84107-25G SIALT84107-5G
Triphenylmethyl mercaptan, Sigma-Aldrich®
Triphenylmethyl mercaptan
Vzorec: C₁₉H₁₆S
MDL Number: MFCD00004854
CAS číslo: 3695-77-0
EINECS: 223-020-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR