Tisk…

Triphenylmethyl mercaptan 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: Triphenylmethanethiol

140410250 140411000 140415000
ACRO140410250EA 3330 CZK
ACRO140410250 ACRO140411000 ACRO140415000
Triphenylmethyl mercaptan 97%
Triphenylmethyl mercaptan
Vzorec: C₁₉H₁₆S
MW: 276,40 g/mol
Bod tání: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00004854
CAS číslo: 3695-77-0
EINECS: 223-020-3

Objednat

Výsledky specifikačního testu

Appearance White to cream to light yellow Powder
Infrared spectrum Conforms
Melting point 102 to 109 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR