Tisk…

N,N-Dibenzylhydroxylamine 98%

Dodavatel: Thermo Fisher Scientific
Varování

B22447.06 B22447.14 B22447.22
B22447.06EA 1680 CZK
B22447.06 B22447.14 B22447.22
N,N-Dibenzylhydroxylamine 98%
N,N-Dibenzylhydroxylamine

Purity: 98% CAS No.: 621-07-8 Molecular Formula: C14H15NO Formula Weight: 213.28 Melting Point: 120-125° Beilstein Reference: 978234

Vzorec: C₁₄H₁₅NO
MW: 213.28 g/mol
Bod tání: 120…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00004772
CAS číslo: 621-07-8
EINECS: 210-667-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR