Tisk…

Salicylohydroxamic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N,2-Dihydroxybenzamide, Salicylhydroxamic acid

M-Clarity™ quality level = MQ200

S607-25G S607-5G
SIALS607-25GEA 3130 CZK
SIALS607-25G SIALS607-5G
Salicylohydroxamic acid, Sigma-Aldrich®
Salicylohydroxamic acid
Vzorec: C₇H₇NO₃
MW: 153,14 g/mol
MDL Number: MFCD00002110
CAS číslo: 89-73-6
EINECS: 201-934-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR