Salicylohydroxamic acid ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
N,2-Dihydroxybenzamide, Salicylhydroxamic acid
A11528.09
A11528.14
A11528.18
A11528.22
A11528.09EA
1300
CZK
A11528.09
A11528.14
A11528.18
A11528.22
Salicylohydroxamic acid ≥99%
Salicylohydroxamic acid
Purity: 99%
CAS No.: 89-73-6
Molecular Formula: C7H7NO3
Formula Weight: 153.14
Melting Point: ca 170° dec.
Beilstein Reference: 1210520
Merck Reference: 14,8331
MDL No.: MFCD00002110
Vzorec:
C₇H₇NO₃ MW: 153,14 g/mol Bod tání: ∼ 170 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00002110 CAS číslo: 89-73-6 EINECS: 201-934-3 Merck Index: 14,8331 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...