Salicylohydroxamic acid 99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
N,2-Dihydroxybenzamide, Salicylhydroxamic acid
132620250
132620050
132620250.EA
3070
CZK
132620250.
132620050.
Salicylohydroxamic acid 99%
Salicylohydroxamic acid
Vzorec:
C₇H₇NO₃ MW: 153,14 g/mol Bod tání: 177…185 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00002110 CAS číslo: 89-73-6 EINECS: 201-934-3 |
Výsledky specifikačního testu
Appearance | Pink-beige Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Elemental Analysis | 8.67 to 9.11 % N |
Specific optical rotation | +6.5° to +9.5° (20°C, 589 nm) c=5 in 1N HCl |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...