Tisk…

Ethyl-4-isocyanatobenzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-(Ethoxycarbonyl)phenyl isocyanate

M-Clarity™ quality level = MQ100

159344-25G 159344-5G
SIAL159344-25GEA 17570 CZK
SIAL159344-25G SIAL159344-5G
Ethyl-4-isocyanatobenzoate, Sigma-Aldrich®
Ethyl-4-isocyanatobenzoate
Vzorec: C₁₀H₉NO₃
MDL Number: MFCD00002027
CAS číslo: 30806-83-8
EINECS: 250-344-2
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR