3-Amino-4-chloro-N,N-dimethylbenzamide 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-Amino-4-chlorobenzamide
B25294.06
B25294.14
B25294.22
B25294.06EA
773
CZK
B25294.06
B25294.14
B25294.22
3-Amino-4-chloro-N,N-dimethylbenzamide 97%
3-Amino-4-chloro-N,N-dimethylbenzamide
Purity: 97%
CAS No.: 19694-10-1
Molecular Formula: C7H7ClN2O
Formula Weight: 170.60
Melting Point: 164-166°
MDL No.: MFCD00035785
Vzorec:
C₇H₇ClN₂O MW: 170,60 g/mol Bod tání: 164…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035785 CAS číslo: 19694-10-1 EINECS: 243-231-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...