Tisk…

Allyl mercaptan, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

06030-100ML 06030-25ML
SIAL06030-100MLEA 11510 CZK
SIAL06030-100ML SIAL06030-25ML
Allyl mercaptan, Sigma-Aldrich®
Allyl mercaptan
Vzorec: C₃H₆S
MDL Number: MFCD00004894
CAS číslo: 870-23-5
EINECS: 212-792-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR