Ethyl-4-nitrophenylglyoxylate ≥98%
Dodavatel: Thermo Scientific
Synonyms:
Ethyl 2-(4-nitrophenyl)-2-oxoacetate
L07579.03
L07579.06
L07579.03EA
2150
CZK
L07579.03
L07579.06
Ethyl-4-nitrophenylglyoxylate ≥98%
Ethyl-4-nitrophenylglyoxylate
Purity: 98+%
CAS No.: 70091-75-7
Molecular Formula: C10H9NO5
Formula Weight: 223.19
Melting Point: 39-43°
Boiling Point: 152-157°/0.5mm
MDL No.: MFCD00051841
Vzorec:
C₁₀H₉NO₅ MW: 223,19 g/mol Bod varu: 152…157 °C (0,5 mmHg) Bod tání: 39…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051841 CAS číslo: 70091-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...