Diethyl (2,4-dichlorophenyl)malonate 95%
Dodavatel: Thermo Scientific
|
Varování
|
H26330.ME
H26330.27
H26330.MEEA
2470
CZK
H26330.ME
H26330.27
Diethyl (2,4-dichlorophenyl)malonate 95%
Diethyl (2,4-dichlorophenyl)malonate
Purity: 95%
CAS No.: 111544-93-5
Molecular Formula: C13H14Cl2O4
Formula Weight: 305.16
MDL No.: MFCD00831103
Vzorec:
C₁₃H₁₄Cl₂O₄ MW: 305,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00831103 CAS číslo: 111544-93-5 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...