Tisk…

7-Hydroxycitronellal, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W258318-10KG W258318-1KG W258318-5KG W258318-SAMPLE
SIALW258318-10KGEA 30880 CZK
SIALW258318-10KG SIALW258318-1KG SIALW258318-5KG SIALW258318-SAMPLE
7-Hydroxycitronellal, Sigma-Aldrich®
7-Hydroxycitronellal
Vzorec: C₁₀H₂₀O₂
MDL Number: MFCD00014681
CAS číslo: 107-75-5
EINECS: 203-518-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR