Tisk…

Aminoiminomethanesulphinic acid 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Thiourea dioxide, FormamidinesulphInic acid

119720100 119721000 119725000
ACRO119720100EA 824 CZK
ACRO119720100 ACRO119721000 ACRO119725000
Aminoiminomethanesulphinic acid 99%
Aminoiminomethanesulphinic acid
Vzorec: CH₄N₂O₂S
MW: 108.12 g/mol
Bod tání: 126 °C (rozkl.)
Storage Temperature: Refrigerator
MDL Number: MFCD00002397
CAS číslo: 1758-73-2
EINECS: 217-157-8
UN: 3341
ADR: 4.2 II

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤0.5 %
Titration with NaOH ≥98.5 %
Impurity ≤0.2 % Thiourea


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR