Tisk…

Acetohydroxamic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N-Hydroxyacetamide

M-Clarity™ quality level = MQ100

159034-10G 159034-50G 159034-5G
SIAL159034-10GEA 4410 CZK
SIAL159034-10G SIAL159034-50G SIAL159034-5G
Acetohydroxamic acid, Sigma-Aldrich®
Acetohydroxamic acid
Vzorec: C₂H₅NO₂
MDL Number: MFCD00009994
CAS číslo: 546-88-3
EINECS: 208-913-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR